Assay Details
| Gene Symbol: APOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003925.1 | NP_001003925.1 | 2 | 746 | 103 | |
| GenBank mRNA | AK232481.1 | - | 2 | 745 | 103 | |
| AK233564.1 | - | 1 | 890 | 103 | ||
| AY583018.1 | - | 2 | 746 | 103 | ||
More Information
Set Membership: |
|

APOF
Genomic Map