Assay Details
Gene Symbol: RNASE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007111.1 | NP_001007112.1 | 2 | 550 | 93 | |
XM_005656307.3 | XP_005656364.1 | 2075 | 93 | |||
XM_013989060.2 | XP_013844514.1 | 2137 | 93 | |||
XM_013989061.2 | XP_013844515.1 | 2083 | 93 | |||
XM_013989063.2 | XP_013844517.1 | 644 | 93 | |||
GenBank mRNA | AJ430467.1 | - | 2 | 550 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
