Assay Details
| Gene Symbol: KLF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011504.1 | NP_001011504.1 | 2 | 650 | 62 | |
| GenBank mRNA | AY850383.1 | - | 2 | 650 | 62 | |
| GQ274320.1 | - | 2 | 1215 | 62 | ||
More Information
Set Membership: |
|

KLF9
Genomic Map