Assay Details
| Gene Symbol: DIRAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044598.2 | NP_001038063.1 | 2 | 452 | 103 | |
| GenBank mRNA | AK347954.1 | - | 2 | 295 | 103 | |
| AK392466.1 | - | 2 | 471 | 103 | ||
| AK397164.1 | - | 1 | 194 | 103 | ||
| DQ529283.1 | - | 2 | 452 | 103 | ||
More Information
Set Membership: |
|

DIRAS3
Genomic Map