Assay Details
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110418.1 | NP_001103888.1 | 2 | 1845 | 104 | |
| XM_021088698.1 | XP_020944357.1 | 1988 | 104 | |||
| XM_021088699.1 | XP_020944358.1 | 2121 | 104 | |||
| XM_021088701.1 | XP_020944360.1 | 1988 | 104 | |||
| XM_021088702.1 | XP_020944361.1 | 2116 | 104 | |||
| XM_021088703.1 | XP_020944362.1 | 2023 | 104 | |||
| XM_021088704.1 | XP_020944363.1 | 1903 | 104 | |||
| XM_021088705.1 | XP_020944364.1 | 2059 | 104 | |||
| GenBank mRNA | DQ005441.1 | - | 2 | 1845 | 104 | |
More Information
Set Membership: |
|

KCNA2
Genomic Map