Assay Details
| Gene Symbol: CMKLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123100.1 | NP_001116572.1 | 4 | 1346 | 76 | |
| XM_021072208.1 | XP_020927867.1 | 1382 | 76 | |||
| XM_021072209.1 | XP_020927868.1 | 1396 | 76 | |||
| GenBank mRNA | AB112778.1 | - | 1 | 677 | 76 | |
| EU421950.1 | - | 4 | 1346 | 76 | ||
More Information
Set Membership: |
|

CMKLR1
Genomic Map