Assay Details
| Gene Symbol: APEX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139471.1 | NP_001132943.1 | 4 | 951 | 96 | |
| XM_021098038.1 | XP_020953697.1 | 1580 | 96 | |||
| XM_021098039.1 | XP_020953698.1 | 1064 | 96 | |||
| GenBank mRNA | AK239484.1 | - | 5 | 939 | 96 | |
| AK389218.1 | - | 5 | 939 | 96 | ||
| FJ358603.1 | - | 4 | 951 | 96 | ||
More Information
Set Membership: |
|

APEX1
Genomic Map