Assay Details
| Gene Symbol: LGALS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142827.1 | NP_001136299.1 | 8 | 744 | 99 | |
| XM_021071980.1 | XP_020927639.1 | 1465 | 99 | |||
| XM_021071981.1 | XP_020927640.1 | 1527 | 99 | |||
| XM_021071982.1 | XP_020927641.1 | 1586 | 99 | |||
| XM_021071983.1 | XP_020927642.1 | 1608 | 99 | |||
| XM_021071984.1 | XP_020927643.1 | 1582 | 99 | |||
| XM_021071985.1 | XP_020927644.1 | 1408 | 99 | |||
| XM_021071986.1 | XP_020927645.1 | 1408 | 99 | |||
| XM_021071987.1 | XP_020927646.1 | 1348 | 99 | |||
| XM_021071988.1 | XP_020927647.1 | 1410 | 99 | |||
| XM_021071989.1 | XP_020927648.1 | 1491 | 99 | |||
| XM_021071990.1 | XP_020927649.1 | 1410 | 99 | |||
| XM_021071991.1 | XP_020927650.1 | 1521 | 99 | |||
| XM_021071992.1 | XP_020927651.1 | 1291 | 99 | |||
| XM_021071993.1 | XP_020927652.1 | 1431 | 99 | |||
| XM_021071994.1 | XP_020927653.1 | 1353 | 99 | |||
| XM_021071995.1 | XP_020927654.1 | 1407 | 99 | |||
| GenBank mRNA | AK344441.1 | - | 9 | 1311 | 99 | |
| FJ424706.1 | - | 8 | 744 | 99 | ||
More Information
Set Membership: |
|

LGALS8
Genomic Map