Assay Details
| Gene Symbol: LCLAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142845.1 | NP_001136317.1 | 6 | 751 | 101 | |
| GenBank mRNA | AK234842.1 | - | 6 | 865 | 101 | |
| AK236662.1 | - | 7 | 967 | 101 | ||
| AK398269.1 | - | 6 | 885 | 101 | ||
| AK400629.1 | - | 6 | 890 | 101 | ||
| FJ424708.1 | - | 6 | 751 | 101 | ||
More Information
Set Membership: |
|

LCLAT1
Genomic Map