Assay Details
| Gene Symbol: GZMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143710.1 | NP_001137182.1 | 4 | 556 | 102 | |
| GenBank mRNA | AK231164.1 | - | 4 | 646 | 102 | |
| AK345183.1 | - | 608 | 102 | |||
| FJ439672.1 | - | 4 | 556 | 102 | ||
More Information
Set Membership: |
|

GZMB
Genomic Map