Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144111.1 | NP_001137583.1 | 2 | 647 | 95 | |
XM_021083799.1 | XP_020939458.1 | 698 | 95 | |||
GenBank mRNA | AK398355.1 | - | 2 | 654 | 95 | |
EU334865.1 | - | 2 | 647 | 95 | ||
EU334866.1 | - | 1 | 384 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
