Assay Details
| Gene Symbol: GPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145019.1 | NP_001138491.1 | 12 | 1450 | 95 | |
| GenBank mRNA | AK233092.1 | - | 12 | 1482 | 95 | |
| AK233431.1 | - | 12 | 1430 | 95 | ||
| AK393113.1 | - | 14 | 2425 | 95 | ||
| FJ439669.1 | - | 12 | 1450 | 95 | ||
| FJ588002.1 | - | 13 | 2367 | 95 | ||
More Information
Set Membership: |
|

GPAT4
Genomic Map