Assay Details
| Gene Symbol: SLC22A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145752.1 | NP_001139224.1 | 7 | 1147 | 87 | |
| XM_005661640.3 | XP_005661697.1 | 1413 | 87 | |||
| XM_021077199.1 | XP_020932858.1 | 1258 | 87 | |||
| GenBank mRNA | FN178491.1 | - | 7 | 1147 | 87 | |
More Information
Set Membership: |
|

SLC22A4
Genomic Map