Assay Details
| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146130.2 | NP_001139602.2 | 4 | 655 | 88 | |
| GenBank mRNA | AK344493.1 | - | 4 | 603 | 88 | |
| FJ713408.1 | - | 4 | 593 | 88 | ||
| FJ713409.1 | - | 5 | 949 | 88 | ||
More Information
Set Membership: |
|

HPSE
Genomic Map