Assay Details
| Gene Symbol: PDK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159306.1 | NP_001152778.1 | 4 | 656 | 103 | |
| GenBank mRNA | EU583382.1 | - | 4 | 656 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PDK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159306.1 | NP_001152778.1 | 4 | 656 | 103 | |
| GenBank mRNA | EU583382.1 | - | 4 | 656 | 103 | |
Set Membership: |
|