Assay Details
| Gene Symbol: MIR9-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038511.1 | 1 | 38 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 161B | TMEM161B | -375.8kb | Sense | Chr. 2 - 95625444 - 95700575 on NCBI Build Sscrofa11.1 | |
| Downstream | myocyte enhancer factor 2C | MEF2C | 46.0kb | Sense | Chr. 2 - 96122407 - 96296927 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR9-2
Genomic Map