Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129969.1 | NP_001123441.1 | 1 - 2 | 160 | 96 | |
GenBank mRNA | AF371322.1 | - | 1 - 2 | 160 | 96 | |
AK390435.1 | - | 1 - 2 | 306 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
