Assay Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163997.1 | NP_001157469.1 | 2 - 3 | 59 | 88 | |
| XM_005655587.3 | XP_005655644.1 | 158 | 88 | |||
| XM_005655589.3 | XP_005655646.1 | 158 | 88 | |||
| GenBank mRNA | AK233447.1 | - | 2 - 3 | 59 | 88 | |
| AK238248.1 | - | 2 - 3 | 58 | 88 | ||
| AK240360.1 | - | 2 - 3 | 59 | 88 | ||
| AK343746.1 | - | 2 - 3 | 60 | 88 | ||
| AK391962.1 | - | 2 - 3 | 66 | 88 | ||
| AK392800.1 | - | 2 - 3 | 72 | 88 | ||
| AK397509.1 | - | 2 - 3 | 60 | 88 | ||
| AK397762.1 | - | 2 - 3 | 73 | 88 | ||
| AK399933.1 | - | 2 - 3 | 72 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163997.1, XM_005655587.3, XM_005655589.3 transcripts.
Set Membership: |
|

MYL6
Genomic Map