Assay Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163997.1 | NP_001157469.1 | 3 - 4 | 206 | 73 | |
| XM_005655587.3 | XP_005655644.1 | 305 | 73 | |||
| XM_005655589.3 | XP_005655646.1 | 305 | 73 | |||
| XM_021091112.1 | XP_020946771.1 | 255 | 73 | |||
| GenBank mRNA | AK230643.1 | - | 3 - 4 | 208 | 73 | |
| AK233447.1 | - | 3 - 4 | 206 | 73 | ||
| AK238248.1 | - | 3 - 4 | 205 | 73 | ||
| AK240054.1 | - | 3 - 4 | 218 | 73 | ||
| AK240358.1 | - | 2 - 3 | 477 | 73 | ||
| AK240360.1 | - | 3 - 4 | 206 | 73 | ||
| AK343746.1 | - | 3 - 4 | 207 | 73 | ||
| AK343941.1 | - | 3 - 4 | 219 | 73 | ||
| AK391962.1 | - | 3 - 4 | 213 | 73 | ||
| AK392800.1 | - | 3 - 4 | 219 | 73 | ||
| AK397509.1 | - | 3 - 4 | 207 | 73 | ||
| AK397762.1 | - | 3 - 4 | 220 | 73 | ||
| AK399933.1 | - | 3 - 4 | 219 | 73 | ||
More Information
Set Membership: |
|

MYL6
Genomic Map