Assay Details
| Gene Symbol: SLC25A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164512.2 | NP_001157984.1 | 2 - 3 | 284 | 80 | |
| GenBank mRNA | AK234146.1 | - | 2 - 3 | 243 | 80 | |
| AK239514.1 | - | 2 - 3 | 214 | 80 | ||
| AK346660.1 | - | 2 - 3 | 214 | 80 | ||
| AK390363.1 | - | 2 - 3 | 284 | 80 | ||
| AK392459.1 | - | 2 - 3 | 215 | 80 | ||
| AK398777.1 | - | 2 - 3 | 255 | 80 | ||
| AK398909.1 | - | 2 - 3 | 240 | 80 | ||
| GQ368242.1 | - | 1 - 2 | 155 | 80 | ||
More Information
Set Membership: |
|

SLC25A3
Genomic Map