Assay Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164513.1 | NP_001157985.1 | 5 - 6 | 617 | 96 | |
| XM_005653912.3 | XP_005653969.1 | 897 | 96 | |||
| XM_005653913.3 | XP_005653970.1 | 868 | 96 | |||
| XM_013979545.2 | XP_013834999.1 | 766 | 96 | |||
| GenBank mRNA | GQ369764.1 | - | 6 - 7 | 836 | 96 | |
| GQ369765.1 | - | 5 - 6 | 617 | 96 | ||
More Information
Set Membership: |
|

ZPBP2
Genomic Map