Assay Details
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166229.1 | NP_001159701.1 | 7 - 8 | 1228 | 65 | |
GenBank mRNA | AF202971.1 | - | 2 - 3 | 253 | 65 | |
AK231224.1 | - | 7 - 8 | 1372 | 65 | ||
AK391240.1 | - | 7 - 8 | 1104 | 65 | ||
EU722905.1 | - | 7 - 8 | 1228 | 65 | ||
X54724.1 | - | 966 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
