Assay Details
| Gene Symbol: ACAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167638.1 | NP_001161110.1 | 7 - 8 | 884 | 71 | |
| GenBank mRNA | AB527054.1 | - | 7 - 8 | 884 | 71 | |
| AK232273.1 | - | 7 - 8 | 912 | 71 | ||
| AK344871.1 | - | 7 - 8 | 1307 | 71 | ||
More Information
Set Membership: |
|

ACAA2
Genomic Map