Assay Details
| Gene Symbol: NDUFAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167652.1 | NP_001161124.1 | 3 - 4 | 379 | 91 | |
| XM_005656762.3 | XP_005656819.2 | 437 | 91 | |||
| GenBank mRNA | AB529863.1 | - | 3 - 4 | 379 | 91 | |
| AK236752.1 | - | 3 - 4 | 432 | 91 | ||
| AK352251.1 | - | 3 - 4 | 400 | 91 | ||
| AK391639.1 | - | 3 - 4 | 443 | 91 | ||
More Information
Set Membership: |
|

NDUFAB1
Genomic Map