Assay Details
| Gene Symbol: ACTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167795.1 | NP_001161267.1 | 1 - 2 | 81 | 99 | |
| XM_021071931.1 | XP_020927590.1 | 103 | 99 | |||
| GenBank mRNA | AK394681.1 | - | 1 - 2 | 79 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001167795.1, XM_021071931.1 transcripts.
Set Membership: |
|

ACTA1
Genomic Map