Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044610.2 | NP_001038075.1 | 1 - 2 | 152 | 91 | |
| GenBank mRNA | AK350066.1 | - | 1 - 2 | 154 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001044610.2 transcripts.
Set Membership: |
|

NME2
Genomic Map