Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044610.2 | NP_001038075.1 | 4 - 5 | 501 | 107 | |
| GenBank mRNA | AB292846.1 | - | 3 - 4 | 359 | 107 | |
| AK350066.1 | - | 4 - 5 | 503 | 107 | ||
| DQ372081.1 | - | 3 - 4 | 345 | 107 | ||
More Information
Set Membership: |
|

NME2
Genomic Map