Assay Details
| Gene Symbol: C7H6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128447.2 | NP_001121919.2 | 1 - 2 | 81 | 79 | |
| GenBank mRNA | HX231531.1 | - | 1 - 2 | 81 | 79 | |
More Information
Set Membership: |
|

C7H6orf15
Genomic Map