Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128472.1 | NP_001121944.1 | 4 - 5 | 539 | 64 | |
| GenBank mRNA | AK347456.1 | - | 4 - 5 | 572 | 64 | |
| AK347693.1 | - | 4 - 5 | 568 | 64 | ||
| EF581989.1 | - | 4 - 5 | 539 | 64 | ||
| EU429459.1 | - | 4 - 5 | 1467 | 64 | ||
More Information
Set Membership: |
|

HSD17B1
Genomic Map