Assay Details
| Gene Symbol: LGALS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142841.1 | NP_001136313.1 | 1 - 2 | 57 | 60 | |
| GenBank mRNA | FJ424703.1 | - | 1 - 2 | 57 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142841.1 transcripts.
Set Membership: |
|

LGALS13
Genomic Map