Assay Details
Gene Symbol: DKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145384.1 | NP_001138856.1 | 1 - 2 | 235 | 85 | |
GenBank mRNA | JN966758.1 | - | 1 - 2 | 405 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
