Assay Details
Gene Symbol: RIIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159765.1 | NP_001153237.1 | 3 - 4 | 213 | 72 | |
XM_013997138.2 | XP_013852592.1 | 252 | 72 | |||
GenBank mRNA | EW576675.2 | - | 3 - 4 | 191 | 72 | |
FD590890.1 | - | 3 - 4 | 215 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
