Assay Details
| Gene Symbol: GUCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160275.1 | NP_001153747.1 | 2 - 3 | 336 | 65 | |
| XM_021093265.1 | XP_020948924.1 | 342 | 65 | |||
| GenBank mRNA | Z83746.1 | - | 336 | 65 | ||
More Information
Set Membership: |
|

GUCA2B
Genomic Map