Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160426.1 | NP_001153898.1 | 3 - 4 | 376 | 63 | |
| XM_021090975.1 | XP_020946634.1 | 721 | 63 | |||
| GenBank mRNA | AK346946.1 | - | 4 - 5 | 630 | 63 | |
| AK390432.1 | - | 4 - 5 | 648 | 63 | ||
More Information
Set Membership: |
|

ARF3
Genomic Map