Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161640.1 | NP_001155112.1 | 8 - 9 | 1193 | 65 | |
| XM_021074526.1 | XP_020930185.1 | 65 | ||||
| GenBank mRNA | AB499815.1 | - | 8 - 9 | 1193 | 65 | |
More Information
Set Membership: |
|

CASP10
Genomic Map