Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170516.1 | NP_001163987.1 | 1 - 2 | 59 | 68 | |
| GenBank mRNA | AK345138.1 | - | 1 - 2 | 192 | 68 | |
| AK346812.1 | - | 1 - 2 | 213 | 68 | ||
| AK346914.1 | - | 1 - 2 | 188 | 68 | ||
| AK347081.1 | - | 1 - 2 | 188 | 68 | ||
| AM911345.1 | - | 1 - 2 | 59 | 68 | ||
More Information
Set Membership: |
|

CCL19
Genomic Map