Assay Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171752.1 | NP_001165223.1 | 6 - 7 | 1546 | 64 | |
XM_005659322.3 | XP_005659379.1 | 1733 | 64 | |||
XM_005659323.3 | XP_005659380.1 | 1695 | 64 | |||
XM_013992715.2 | XP_013848169.1 | 1754 | 64 | |||
XM_013992716.2 | XP_013848170.1 | 1590 | 64 | |||
XM_013992717.2 | XP_013848171.1 | 1715 | 64 | |||
GenBank mRNA | FJ719777.1 | - | 6 - 7 | 1546 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
