Assay Details
Gene Symbol: SKP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171755.1 | NP_001165226.1 | 5 - 6 | 672 | 59 | |
GenBank mRNA | AK395880.1 | - | 5 - 6 | 854 | 59 | |
FJ536864.1 | - | 5 - 6 | 672 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
