Assay Details
| Gene Symbol: APOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174132.2 | NP_001167603.1 | 1 - 2 | 106 | 114 | |
| GenBank mRNA | GU386346.2 | - | 1 - 2 | 106 | 114 | |
More Information
Set Membership: |
|

APOC4
Genomic Map