Assay Details
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185049.2 | NP_001171978.2 | 6 - 7 | 2572 | 96 | |
XM_021100320.1 | XP_020955979.1 | 2596 | 96 | |||
XM_021100322.1 | XP_020955981.1 | 2515 | 96 | |||
XM_021100323.1 | XP_020955982.1 | 2506 | 96 | |||
XM_021100324.1 | XP_020955983.1 | 2491 | 96 | |||
XM_021100325.1 | XP_020955984.1 | 2482 | 96 | |||
XM_021100326.1 | XP_020955985.1 | 1014 | 96 | |||
XM_021100327.1 | XP_020955986.1 | 4031 | 96 | |||
GenBank mRNA | HM037344.1 | - | 5 - 6 | 637 | 96 | |
HM037345.1 | - | 5 - 6 | 628 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
