Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185065.1 | NP_001171994.1 | 2 - 3 | 286 | 75 | |
| XM_021088493.1 | XP_020944152.1 | 632 | 75 | |||
| GenBank mRNA | AY610482.1 | - | 2 - 3 | 291 | 75 | |
More Information
Set Membership: |
|

PMP2
Genomic Map