Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185067.1 | NP_001171996.1 | 1 - 2 | 218 | 67 | |
| XM_021097975.1 | XP_020953634.1 | 448 | 67 | |||
| XM_021097976.1 | XP_020953635.1 | 337 | 67 | |||
| GenBank mRNA | AK236491.1 | - | 1 - 2 | 163 | 67 | |
| AY609861.1 | - | 1 - 2 | 218 | 67 | ||
More Information
Set Membership: |
|

MRPL2
Genomic Map