Assay Details
| Gene Symbol: MRPL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185151.1 | NP_001172080.1 | 3 - 4 | 177 | 69 | |
| XM_005666270.3 | XP_005666327.1 | 191 | 69 | |||
| XM_021098131.1 | XP_020953790.1 | 209 | 69 | |||
| GenBank mRNA | AK238583.1 | - | 3 - 4 | 194 | 69 | |
| AK392634.1 | - | 3 - 4 | 169 | 69 | ||
| AY609459.1 | - | 3 - 4 | 162 | 69 | ||
More Information
Set Membership: |
|

MRPL52
Genomic Map