Assay Details
| Gene Symbol: ARL6IP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185160.1 | NP_001172089.1 | 1 - 2 | 140 | 84 | |
| NM_001185161.1 | NP_001172090.1 | 1 - 2 | 140 | 84 | ||
| XM_021072018.1 | XP_020927677.1 | 162 | 84 | |||
| XM_021072019.1 | XP_020927678.1 | 162 | 84 | |||
| GenBank mRNA | AK240231.1 | - | 1 - 2 | 139 | 84 | |
| BP141292.1 | - | 1 - 2 | 129 | 84 | ||
| CJ008052.1 | - | 1 - 2 | 140 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185160.1, NM_001185161.1, XM_021072018.1, XM_021072019.1 transcripts.
Set Membership: |
|

ARL6IP4
Genomic Map