Assay Details
| Gene Symbol: ARL6IP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185160.1 | NP_001172089.1 | 3 - 4 | 616 | 54 | |
| NM_001185161.1 | NP_001172090.1 | 3 - 4 | 592 | 54 | ||
| NR_033706.1 | 2 - 3 | 874 | 54 | |||
| XM_021072017.1 | XP_020927676.1 | 605 | 54 | |||
| XM_021072018.1 | XP_020927677.1 | 638 | 54 | |||
| XM_021072019.1 | XP_020927678.1 | 614 | 54 | |||
| GenBank mRNA | AK231302.1 | - | 2 - 3 | 862 | 54 | |
| AK240231.1 | - | 3 - 4 | 591 | 54 | ||
| AK393400.1 | - | 2 - 3 | 947 | 54 | ||
| BP141292.1 | - | 3 - 4 | 605 | 54 | ||
| CJ008052.1 | - | 3 - 4 | 592 | 54 | ||
More Information
Set Membership: |
|

ARL6IP4
Genomic Map