Assay Details
| Gene Symbol: ARL6IP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185160.1 | NP_001172089.1 | 5 - 6 | 809 | 72 | |
| NM_001185161.1 | NP_001172090.1 | 5 - 6 | 785 | 72 | ||
| XM_021072017.1 | XP_020927676.1 | 798 | 72 | |||
| GenBank mRNA | AK240231.1 | - | 5 - 6 | 784 | 72 | |
| AK393400.1 | - | 4 - 5 | 1140 | 72 | ||
| BP141292.1 | - | 5 - 6 | 798 | 72 | ||
More Information
Set Membership: |
|

ARL6IP4
Genomic Map