Assay Details
Gene Symbol: MRPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185168.1 | NP_001172097.1 | 2 - 3 | 409 | 92 | |
XM_021088528.1 | XP_020944187.1 | 92 | ||||
GenBank mRNA | AY609523.1 | - | 2 - 3 | 409 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
