Assay Details
| Gene Symbol: UQCRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185172.1 | NP_001172101.1 | 1 - 2 | 56 | 72 | |
| GenBank mRNA | EV897222.2 | - | 1 - 2 | 65 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185172.1 transcripts.
Set Membership: |
|

UQCRB
Genomic Map