Assay Details
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185174.1 | NP_001172103.1 | 1 - 2 | 92 | 93 | |
GenBank mRNA | AY609391.1 | - | 1 - 2 | 93 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
