Assay Details
| Gene Symbol: SLC25A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190189.1 | NP_001177118.1 | 4 - 5 | 556 | 68 | |
| XM_021071932.1 | XP_020927591.1 | 848 | 68 | |||
| GenBank mRNA | AK234877.1 | - | 4 - 5 | 556 | 68 | |
More Information
Set Membership: |
|

SLC25A1
Genomic Map